ENA423199965 Enamine

1-[1-(ethanesulfonyl)azetidin-3-yl]methanamine hydrochloride

  • MDL number MFCD27959457
  • Cas Number : 1803590-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160385
Availability details for
Quantity
  • Availability
  • Add to Cart