ENA423199967 Enamine

2-(ethylsulfamoyl)-4-nitrobenzoic acid

  • MDL number MFCD27980730
  • Cas Number : 1803566-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160411
Availability details for
Quantity
  • Availability
  • Add to Cart