ENA423199972 Enamine

1-{6-[(2,2,2-trifluoroethoxy)methyl]pyridin-3-yl}methanamine dihydrochloride

  • MDL number MFCD27959464
  • Cas Number : 1803588-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160509
Availability details for
Quantity
  • Availability
  • Add to Cart