ENA423199975 Enamine

methyl 1-benzyl-1H-1,2,3-triazole-4-carboxylate

  • MDL number MFCD03084464
  • Cas Number : 76003-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141543
Availability details for
Quantity
  • Availability
  • Add to Cart