ENA423199996 Enamine

3-hydroxycyclohexane-1-carboxylic acid

  • MDL number MFCD19228828
  • Cas Number : 606488-94-2
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-hydroxycyclohexanecarboxylic acid
Inchi Code 1S/C7H12O3/c8-6-3-1-2-5(4-6)7(9)10/h5-6,8H,1-4H2,(H,9,10)
Inchi Key JBZDHFKPEDWWJC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-151572

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-151572
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart