ENA423200001 Enamine

2-tert-butyl-6-(propan-2-yl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD12137541
  • Cas Number : 1153296-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152324
Availability details for
Quantity
  • Availability
  • Add to Cart