ENA423200022 Enamine

1H-imidazol-2-yl(pyridin-3-yl)methanol

  • MDL number MFCD23866516
  • Cas Number : 1518433-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153282

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153282
Availability details for
Quantity
  • Availability
  • Add to Cart