ENA423200033 Enamine

6-chloro-2-(trifluoromethyl)pyridine-3-carbaldehyde

  • MDL number MFCD16610713
  • Cas Number : 1227581-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153610

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153610
Availability details for
Quantity
  • Availability
  • Add to Cart