ENA423200041 Enamine

1-(2,5-diethylpiperazin-1-yl)-2-methylpropan-2-ol

  • MDL number MFCD21118445
  • Cas Number : 1492369-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154161
Availability details for
Quantity
  • Availability
  • Add to Cart