ENA423200042 Enamine

2-{[(1-hydroxy-3-methylcyclohexyl)methyl]amino}-N-methylacetamide

  • MDL number MFCD21165292
  • Cas Number : 1477679-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154164
Availability details for
Quantity
  • Availability
  • Add to Cart