ENA423200043 Enamine

1-(2-methoxyethyl)-1,4-diazepan-2-one

  • MDL number MFCD21195081
  • Cas Number : 1513297-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154168
Availability details for
Quantity
  • Availability
  • Add to Cart