ENA423200049 Enamine

1-[(2-hydroxyethyl)amino]cyclohexane-1-carbonitrile

  • MDL number MFCD00506673
  • Cas Number : 54961-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154182
Availability details for
Quantity
  • Availability
  • Add to Cart