ENA423200062 Enamine

tert-butyl 4-(2-bromoethyl)piperazine-1-carboxylate

  • MDL number MFCD07367890
  • Cas Number : 655225-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154245

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-154245
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart