ENA423200069 Enamine

tert-butyl (2S)-2-(hydroxymethyl)piperidine-1-carboxylate

  • MDL number MFCD04973121
  • Cas Number : 134441-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81-84
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-154305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154305
Availability details for
Quantity
  • Availability
  • Add to Cart