ENA423200070 Enamine

2,8-diazaspiro[5.5]undecan-1-one hydrochloride

  • MDL number MFCD08461170
  • Cas Number : 1181265-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154316

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154316
Availability details for
Quantity
  • Availability
  • Add to Cart