ENA423200081 Enamine

2-(2-methoxyethyl)-2-methylbut-3-enoic acid

  • MDL number MFCD27959358
  • Cas Number : 1596663-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154718

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154718
Availability details for
Quantity
  • Availability
  • Add to Cart