ENA423200085 Enamine

1-methyl-1H-1,2,3-triazole-5-sulfonamide

  • MDL number MFCD27980647
  • Cas Number : 1602410-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154827
Availability details for
Quantity
  • Availability
  • Add to Cart