ENA423200087 Enamine

4-chloro-6-hydrazinylpyrimidin-5-amine

  • MDL number MFCD04035542
  • Cas Number : 42786-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154969
Availability details for
Quantity
  • Availability
  • Add to Cart