ENA423200088 Enamine

6-methoxy-1,2-benzothiazole-3-carboxylic acid

  • MDL number MFCD11042715
  • Cas Number : 677304-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155032
Availability details for
Quantity
  • Availability
  • Add to Cart