ENA423200093 Enamine

methyl 2-amino-4,5-dimethyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

  • MDL number MFCD20408787
  • Cas Number : 1343247-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155293
Availability details for
Quantity
  • Availability
  • Add to Cart