ENA423200098 Enamine

(5-chloro-1-methyl-1H-1,3-benzodiazol-2-yl)methanamine

  • MDL number MFCD08572217
  • Cas Number : 937635-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155337

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155337
Availability details for
Quantity
  • Availability
  • Add to Cart