ENA423200102 Enamine

6-bromo-5-chloropyridine-3-carboxylic acid

  • MDL number MFCD03426922
  • Cas Number : 38185-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155406

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155406
Availability details for
Quantity
  • Availability
  • Add to Cart