ENA423200106 Enamine

1-(2-methoxypyridin-4-yl)ethan-1-one

  • MDL number MFCD10697715
  • Cas Number : 764708-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155490

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155490
Availability details for
Quantity
  • Availability
  • Add to Cart