ENA423200107 Enamine

2-({1-[(tert-butoxy)carbonyl]-3-methylazetidin-3-yl}oxy)acetic acid

  • MDL number MFCD27959363
  • Cas Number : 1602349-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-155504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155504
Availability details for
Quantity
  • Availability
  • Add to Cart