ENA423200115 Enamine

2-(2,5-dimethylphenyl)-6-fluoro-2,3-dihydro-1,2-benzothiazol-3-one

  • MDL number MFCD05859599
  • Cas Number : 727664-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155714
Availability details for
Quantity
  • Availability
  • Add to Cart