ENA423200116 Enamine

1,2-di-tert-butyl 5-methoxypyrrolidine-1,2-dicarboxylate

  • MDL number MFCD24537134
  • Cas Number : 178961-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155729

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155729
Availability details for
Quantity
  • Availability
  • Add to Cart