ENA423200119 Enamine

4-(benzyloxy)-2-nitroaniline

  • MDL number MFCD00024263
  • Cas Number : 26697-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155777

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155777
Availability details for
Quantity
  • Availability
  • Add to Cart