ENA423200129 Enamine

1-(2-methoxy-5-methylphenyl)ethan-1-ol

  • MDL number MFCD08442415
  • Cas Number : 926255-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156115
Availability details for
Quantity
  • Availability
  • Add to Cart