ENA423200131 Enamine

3-methylisoquinolin-5-amine

  • MDL number MFCD00160674
  • Cas Number : 54410-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115240

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115240
Availability details for
Quantity
  • Availability
  • Add to Cart