ENA423200135 Enamine

4-bromo-1-(bromomethyl)-2-methylbenzene

  • MDL number MFCD12025011
  • Cas Number : 156001-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156205

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156205
Availability details for
Quantity
  • Availability
  • Add to Cart