ENA423200138 Enamine

5-phenyl-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD04186194
  • Cas Number : 13743-14-1
  • Molecular Weight : 205.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 185-186
Iupac Name 5-phenyl-1,3-thiazole-4-carboxylic acid
Inchi Code 1S/C10H7NO2S/c12-10(13)8-9(14-6-11-8)7-4-2-1-3-5-7/h1-6H,(H,12,13)
Inchi Key BIIMZWZUYMBFEV-UHFFFAOYSA-N

Description

Melting Point 185-186
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-156283

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-156283
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart