ENA423200152 Enamine

2-hydroxy-3-(3-methoxyphenyl)propanoic acid

  • MDL number MFCD11935533
  • Cas Number : 1181574-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117595
Availability details for
Quantity
  • Availability
  • Add to Cart