ENA423200153 Enamine

2-bromo-6-cyclopentylimidazo[2,1-b][1,3,4]thiadiazole

  • MDL number MFCD16830416
  • Cas Number : 1247646-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156597
Availability details for
Quantity
  • Availability
  • Add to Cart