ENA423200167 Enamine

5-chloro-2-(cyclohexyloxy)benzene-1-sulfonyl chloride

  • MDL number MFCD17028970
  • Cas Number : 1341440-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156790

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156790
Availability details for
Quantity
  • Availability
  • Add to Cart