ENA423200171 Enamine

3-chloro-4-(propan-2-yl)benzoic acid

  • MDL number MFCD20640283
  • Cas Number : 161622-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156811
Availability details for
Quantity
  • Availability
  • Add to Cart