ENA423200179 Enamine

1-[2-(benzyloxy)-4-methoxyphenyl]ethan-1-ol

  • MDL number MFCD24280107
  • Cas Number : 879487-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157012

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157012
Availability details for
Quantity
  • Availability
  • Add to Cart