ENA423200185 Enamine

1,3,4-oxadiazol-2-amine

  • MDL number MFCD17677516
  • Cas Number : 3775-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101637

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-101637
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart