ENA423200189 Enamine

4-(2-bromophenyl)piperidine-2,6-dione

  • MDL number MFCD18277017
  • Cas Number : 99983-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157102
Availability details for
Quantity
  • Availability
  • Add to Cart