ENA423200192 Enamine

3-fluoroquinoline

  • MDL number MFCD00234493
  • Cas Number : 396-31-6
  • Molecular Weight : 147.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-118999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118999
Availability details for
Quantity
  • Availability
  • Add to Cart