ENA423200195 Enamine

2-(1H-1,2,3,4-tetrazol-5-yl)acetamide

  • MDL number MFCD05858433
  • Cas Number : 501356-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157190

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157190
Availability details for
Quantity
  • Availability
  • Add to Cart