ENA423200201 Enamine

N-ethyl-5,6,7,8-tetrahydroquinolin-8-amine

  • MDL number MFCD20684873
  • Cas Number : 878025-41-3
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-ethyl-5,6,7,8-tetrahydro-8-quinolinamine
Inchi Code 1S/C11H16N2/c1-2-12-10-7-3-5-9-6-4-8-13-11(9)10/h4,6,8,10,12H,2-3,5,7H2,1H3
Inchi Key NKBKRVWPGIIUGY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-119199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119199
Availability details for
Quantity
  • Availability
  • Add to Cart