ENA423200209 Enamine

4-[(4-methoxyphenyl)sulfanyl]butanoic acid

  • MDL number MFCD09811045
  • Cas Number : 52872-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157393

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157393
Availability details for
Quantity
  • Availability
  • Add to Cart