ENA423200211 Enamine

5H,6H,7H-pyrrolo[3,4-b]pyridin-5-one

  • MDL number MFCD12923779
  • Cas Number : 40107-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204-206
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120438
Availability details for
Quantity
  • Availability
  • Add to Cart