ENA423200214 Enamine

5-(trifluoromethyl)-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD06803666
  • Cas Number : 37461-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-157416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157416
Availability details for
Quantity
  • Availability
  • Add to Cart