ENA423200215 Enamine

1-benzyl-1H-pyrazol-4-ol

  • MDL number MFCD23110926
  • Cas Number : 226989-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157419

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-157419
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart