ENA423200224 Enamine

1-(5-bromothiophen-2-yl)-4-chlorobutan-1-one

  • MDL number MFCD14532152
  • Cas Number : 80775-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157532

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157532
Availability details for
Quantity
  • Availability
  • Add to Cart