ENA423200225 Enamine

ethyl 4,5-dimethyl-2-phenylfuran-3-carboxylate

  • MDL number MFCD27980668
  • Cas Number : 600158-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-157533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157533
Availability details for
Quantity
  • Availability
  • Add to Cart