ENA423200242 Enamine

tert-butyl 3-cyclobutyl-2-methyl-3-oxopropanoate

  • MDL number MFCD21753379
  • Cas Number : 1512484-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157625

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157625
Availability details for
Quantity
  • Availability
  • Add to Cart