ENA423200244 Enamine

2-bromo-5-(4-chlorophenyl)-1,3,4-oxadiazole

  • MDL number MFCD21900634
  • Cas Number : 1368558-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157632
Availability details for
Quantity
  • Availability
  • Add to Cart