ENA423200247 Enamine

(1S)-1-(2-methoxy-4-methylphenyl)ethan-1-ol

  • MDL number MFCD18339558
  • Cas Number : 1344920-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157640

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157640
Availability details for
Quantity
  • Availability
  • Add to Cart